(11bS)-4-Hydroxy-2,6-dimesityldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%

Supplier: Ambeed
Warning

A1006139-100MG A1006139-1G
77619-612EA 132.87 USD
77619-612 77619-614
(11bS)-4-Hydroxy-2,6-dimesityldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%
(11bS)-4-Hydroxy-2,6-dimesityldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₃₈H₃₃O₄P
MW: 584.6400 g/mol
CAS Number: 878111-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR