3,3'-(Propane-2,2-diylbis(sulfanediyl))dipropionic acid ≥96%

Supplier: Ambeed
Warning

A1005397-100MG A1005397-1G A1005397-250MG A1005397-5G
77619-560EA 42.23 USD
77619-560 77619-562 77619-564 77782-774
3,3'-(Propane-2,2-diylbis(sulfanediyl))dipropionic acid ≥96%
3,3'-(Propane-2,2-diylbis(sulfanediyl))dipropionic acid

Precaución: for Research Use Only

Formula: C₉H₁₆O₄S₂
MDL Number: MFCD32068154
CAS Number: 4265-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR