4-(4-Chlorothiophen-2-yl)thiazol-2-amine ≥97%

Supplier: Ambeed
Warning

A1005378-1G A1005378-25G A1005378-5G
77782-771EA 10 USD
77782-771 77782-772 77782-773
4-(4-Chlorothiophen-2-yl)thiazol-2-amine ≥97%
4-(4-Chlorothiophen-2-yl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂S₂
CAS Number: 570407-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR