2,4,8,10-Tetra-tert-butyl-6-hydroxydibenzo[d,f][1,3,2]dioxaphosphepine 6-oxide ≥98%
Supplier: Ambeed
|
Warning
|
A1005326-1G
A1005326-100MG
A1005326-250MG
77595-668EA
925.58
USD
77595-668
77619-556
77619-558
2,4,8,10-Tetra-tert-butyl-6-hydroxydibenzo[d,f][1,3,2]dioxaphosphepine 6-oxide ≥98%
2,4,8,10-Tetra-tert-butyl-6-hydroxydibenzo[d,f][1,3,2]dioxaphosphepine 6-oxide
Precaución: for Research Use Only
Formula:
C₂₈H₄₁O₄P MW: 472.6 g/mol |
MDL Number:
MFCD34597740 CAS Number: 342794-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...