2,2'-Di(pyridin-4-yl)-1H,1'H-5,5'-bibenzo[d]imidazole ≥98%

Supplier: Ambeed
Warning

A1005254-250MG A1005254-100MG A1005254-1G
77595-666EA 326.98 USD
77595-666 77619-552 77782-768
2,2'-Di(pyridin-4-yl)-1H,1'H-5,5'-bibenzo[d]imidazole ≥98%
2,2'-Di(pyridin-4-yl)-1H,1'H-5,5'-bibenzo[d]imidazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆N₆
MDL Number: MFCD28963972
CAS Number: 308362-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR