4-(2-Methoxy-2-oxoethyl)benzoic acid ≥95%

Supplier: Ambeed
Warning

A241650-10G A241650-5G
77672-160EA 310 USD
77672-160 77672-162
4-(2-Methoxy-2-oxoethyl)benzoic acid ≥95%
4-(2-Methoxy-2-oxoethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD18782075
CAS Number: 87524-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR