5'-Fluoro-2'-hydroxy-3'-nitroacetophenone ≥95%

Supplier: Ambeed
Danger

Synonyms: 1-(5-Fluoro-2-hydroxy-3-nitrophenyl)ethanone

A241265-1G A241265-5G
77612-340EA 59.74 USD
77612-340 77685-673
5'-Fluoro-2'-hydroxy-3'-nitroacetophenone ≥95%
5'-Fluoro-2'-hydroxy-3'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MW: 199.136 g/mol
MDL Number: MFCD03094007
CAS Number: 70978-39-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR