Dimethyl 3,3'-disulfanediyl(2R,2'R)-bis(2-((tert-butoxycarbonyl)amino)propanoate) ≥97%

Supplier: Ambeed
Warning

Synonyms: (Boc-Cystine-OMe)₂

A240787-1G A240787-5G
77879-422EA 103 USD
77879-422 77879-423
Dimethyl 3,3'-disulfanediyl(2R,2'R)-bis(2-((tert-butoxycarbonyl)amino)propanoate) ≥97%
Dimethyl 3,3'-disulfanediyl(2R,2'R)-bis(2-((tert-butoxycarbonyl)amino)propanoate)

Precaución: for Research Use Only

Formula: C₁₈H₃₂N₂O₈S₂
MW: 468.59 g/mol
MDL Number: MFCD09757603
CAS Number: 77826-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR