(S)-2-(Prop-2-yn-1-yl)pyrrolidine-2-carboxylic acid hydrochloride ≥95%

Supplier: Ambeed
Warning

A240699-1G A240699-100MG
77612-336EA 293.55 USD
77612-336 77685-670
(S)-2-(Prop-2-yn-1-yl)pyrrolidine-2-carboxylic acid hydrochloride ≥95%
(S)-2-(Prop-2-yn-1-yl)pyrrolidine-2-carboxylic acid hydrochloride
Formula: C₈H₁₂ClNO₂
MDL Number: MFCD06659383
CAS Number: 1217828-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR