(1S)-2,2'-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)-1,1'-binaphthalene ≥97%

Supplier: Ambeed
Warning

A239901-1G A239901-250MG
77748-418EA 692 USD
77748-418 77748-419
(1S)-2,2'-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)-1,1'-binaphthalene ≥97%
(1S)-2,2'-Bis((S)-4-phenyl-4,5-dihydrooxazol-2-yl)-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₃₈H₂₈N₂O₂
MW: 544.64 g/mol
CAS Number: 180981-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR