4-(Dibutylamino)benzaldehyde ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-dibutylamino-benzaldehyde

A239153-100MG A239153-1G A239153-250MG
77685-666EA 35 USD
77685-666 77685-667 77685-668
4-(Dibutylamino)benzaldehyde ≥98%
4-(Dibutylamino)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₂₃NO
MW: 233.355 g/mol
MDL Number: MFCD00075134
CAS Number: 90134-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR