(E)-5-(4-Methoxystyryl)benzene-1,3-diol ≥98%

Supplier: Ambeed
Warning

Synonyms: 4'-Methoxyresveratrol

A238278-10MG A238278-1MG A238278-5MG
77600-912EA 19.48 USD
77600-912 77600-914 77600-916
(E)-5-(4-Methoxystyryl)benzene-1,3-diol ≥98%
(E)-5-(4-Methoxystyryl)benzene-1,3-diol

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.26 g/mol
MDL Number: MFCD12407152
CAS Number: 33626-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR