Methyl 5-bromo-2-fluoro-3-nitrobenzoate ≥95%

Supplier: Ambeed
Warning

A238040-5G A238040-250MG
77612-312EA 82.4 USD
77612-312 77683-710
Methyl 5-bromo-2-fluoro-3-nitrobenzoate ≥95%
Methyl 5-bromo-2-fluoro-3-nitrobenzoate

Precaución: For research use only.

Formula: C₈H₅BrFNO₄
MW: 278.03 g/mol
MDL Number: MFCD12172992
CAS Number: 1154278-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR