5,8-Dioxaspiro[3.4]octane-2-methanol ≥97%

Supplier: Ambeed
Warning

A237191-1G A237191-250MG A237191-5G
77892-408EA 167 USD
77892-408 77892-409 77892-410
5,8-Dioxaspiro[3.4]octane-2-methanol ≥97%
5,8-Dioxaspiro[3.4]octane-2-methanol

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MDL Number: MFCD23106218
CAS Number: 545882-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR