3-Benzylmorpholine hydrochloride ≥98%

Supplier: Ambeed
Warning

A236961-100MG A236961-250MG
77885-604EA 45 USD
77885-604 77885-605
3-Benzylmorpholine hydrochloride ≥98%
3-Benzylmorpholine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO·ClH
MDL Number: MFCD04117744
CAS Number: 1172897-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR