9-(2-Ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98%

Supplier: Ambeed
Warning

Synonyms: 9-(2-Ethylhexyl)-9H-carbazole-2,7-diboronic acid bis(pinacol) ester

A236907-100MG A236907-1G A236907-250MG
77683-705EA 110 USD
77683-705 77683-706 77683-707
9-(2-Ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98%
9-(2-Ethylhexyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole

Precaución: For research use only.

Formula: C₃₂H₄₇B₂NO₄
MW: 531.35 g/mol
MDL Number: MFCD23703117
CAS Number: 476360-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR