(R)-2-(3-((Benzo[d]oxazol-2-yl(3-(4-methoxyphenoxy)propyl)amino)methyl)phenoxy)butanoic acid ≥99%

Supplier: Ambeed
Warning

Synonyms: pemafibrate

A236714-10MG A236714-1MG A236714-5MG
77615-956EA 72.1 USD
77615-956 77615-958 77615-960
(R)-2-(3-((Benzo[d]oxazol-2-yl(3-(4-methoxyphenoxy)propyl)amino)methyl)phenoxy)butanoic acid ≥99%
(R)-2-(3-((Benzo[d]oxazol-2-yl(3-(4-methoxyphenoxy)propyl)amino)methyl)phenoxy)butanoic acid

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₂O₆
MW: 490.55 g/mol
MDL Number: MFCD30533427
CAS Number: 848259-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR