4-Bromo-2-isopropoxypyridine ≥98%

Supplier: Ambeed
Warning

A236624-1G A236624-250MG
77890-371EA 35 USD
77890-371 77890-372
4-Bromo-2-isopropoxypyridine ≥98%
4-Bromo-2-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MDL Number: MFCD11869614
CAS Number: 1142194-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR