Fmoc-Met-Opfp 98+%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Met-OPfp

A236104-1G A236104-5G
77667-942EA 18 USD
77667-942 77879-409
Fmoc-Met-Opfp 98+%
Fmoc-Met-Opfp

Precaución: for Research Use Only

Formula: C₂₆H₂₀F₅NO₄S
MDL Number: MFCD00065665
CAS Number: 86060-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR