Thymidine 5′-triphosphate sodium salt (dTTP Na) ≥97%

Supplier: Ambeed
Warning

Synonyms: Thymidine 5′-(tetrahydrogen triphosphate) xsodium salt, 2'-Deoxythymidine-5'-triphosphoric acid sodium salt, Thymidine-5'-triphosphoric acid sodium salt, TTP Na, dTTP Na, Deoxythymidine triphosphate sodium salt, dTTP sodium salt

A236087-100MG A236087-250MG
77683-700EA 115 USD
77683-700 77683-701
Thymidine 5′-triphosphate sodium salt (dTTP Na) ≥97%
Thymidine 5′-triphosphate sodium salt (dTTP Na)

Precaución: For research use only.

Formula: C₁₀H₁₆N₂NaO₁₄P₃
MW: 504.15 g/mol
MDL Number: MFCD00080338
CAS Number: 18423-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR