(S)-2-(2,4-Dihydroxyphenyl)-7-hydroxy-5-methoxy-8-((R)-5-methyl-2-(prop-1-en-2-yl)hex-4-en-1-yl)chroman-4-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Kurarinone

A236069-1MG A236069-5MG
77667-938EA 39 USD
77667-938 77667-940
(S)-2-(2,4-Dihydroxyphenyl)-7-hydroxy-5-methoxy-8-((R)-5-methyl-2-(prop-1-en-2-yl)hex-4-en-1-yl)chroman-4-one ≥98%
(S)-2-(2,4-Dihydroxyphenyl)-7-hydroxy-5-methoxy-8-((R)-5-methyl-2-(prop-1-en-2-yl)hex-4-en-1-yl)chroman-4-one

Precaución: for Research Use Only

Formula: C₂₆H₃₀O₆
MW: 438.51 g/mol
CAS Number: 34981-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR