(9H-Fluoren-9-yl)methyl 4-aminobenzylcarbamate ≥95%

Supplier: Ambeed
Warning

A235729-1G A235729-250MG A235729-25G A235729-5G
77890-364EA 38 USD
77890-364 77890-365 77890-366 77890-367
(9H-Fluoren-9-yl)methyl 4-aminobenzylcarbamate ≥95%
(9H-Fluoren-9-yl)methyl 4-aminobenzylcarbamate

Precaución: for Research Use Only

Formula: C₂₂H₂₀N₂O₂
MDL Number: MFCD06658452
CAS Number: 159790-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR