(+)-1-Deoxynojirimycin ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Deoxynojirimycin, (2R,3R,4R,5S)-2-(Hydroxymethyl)piperidine-3,4,5-triol

A235571-100MG A235571-1MG A235571-250MG A235571-2MG A235571-5MG
77612-302EA 56.65 USD
77612-302 77685-650 77685-651 77685-652 77685-653
(+)-1-Deoxynojirimycin ≥98%
(+)-1-Deoxynojirimycin

Precaución: for Research Use Only

Formula: C₆H₁₃NO₄
MW: 163.17 g/mol
MDL Number: MFCD00063474
CAS Number: 19130-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR