(Isobuyl)triphenylphosphonium bromide ≥98%

Supplier: Ambeed
Warning

Synonyms: Isobutyltriphenylphosphonium bromide

A235105-1G A235105-5G
77600-890EA 9.23 USD
77600-890 77600-892
(Isobuyl)triphenylphosphonium bromide ≥98%
(Isobuyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: (H₃C)₂HCP(C₆H₅)₃Br
MW: 399.31 g/mol
MDL Number: MFCD00031593
CAS Number: 22884-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR