N-Boc-D-cyclohexylglycinol ≥98%

Supplier: Ambeed
Warning

A234991-1G A234991-5G
77883-619EA 10 USD
77883-619 77883-620
N-Boc-D-cyclohexylglycinol ≥98%
N-Boc-D-cyclohexylglycinol

Precaución: for Research Use Only

Formula: C₁₃H₂₅NO₃
MDL Number: MFCD04112590
CAS Number: 188348-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR