Methyl 4-chloro-5-methylpicolinate ≥97%

Supplier: Ambeed
Warning

A234311-100MG A234311-250MG
77883-615EA 69 USD
77883-615 77883-616
Methyl 4-chloro-5-methylpicolinate ≥97%
Methyl 4-chloro-5-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD16987786
CAS Number: 1104455-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR