2,6-Diiodo-4-(trifluoromethyl)aniline ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A233655-1G A233655-5G
77683-684EA 81 USD
77683-684 77885-594
2,6-Diiodo-4-(trifluoromethyl)aniline ≥95%, Ambeed.Inc
2,6-Diiodo-4-(trifluoromethyl)aniline

Precaución: For research use only.

Formula: C₇H₄F₃I₂N
MW: 412.92 g/mol
MDL Number: MFCD11847372
CAS Number: 214400-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR