Sodium (6R,7S)-7-(2-(((S)-2-amino-2-carboxyethyl)thio)acetamido)-7-methoxy-3-(((1-methyl-1H-tetrazol-5-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate ≥97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Cefminox sodium
A233269-10MG
A233269-1MG
77667-922EA
22
USD
77667-922
77667-924
Sodium (6R,7S)-7-(2-(((S)-2-amino-2-carboxyethyl)thio)acetamido)-7-methoxy-3-(((1-methyl-1H-tetrazol-5-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate ≥97%
Sodium (6R,7S)-7-(2-(((S)-2-amino-2-carboxyethyl)thio)acetamido)-7-methoxy-3-(((1-methyl-1H-tetrazol-5-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₆H₂₀N₇NaO₇S₃ MW: 541.55 g/mol |
MDL Number:
MFCD00941387 CAS Number: 75498-96-3 UN: 1325 ADR: 4.1 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...