2-Fluoro-6-methylbenzoyl chloride ≥98%

Supplier: Ambeed
Danger

A233089-5G A233089-1G
77672-070EA 260 USD
77672-070 77883-613
2-Fluoro-6-methylbenzoyl chloride ≥98%
2-Fluoro-6-methylbenzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MDL Number: MFCD06660278
CAS Number: 535961-78-5
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR