Azure C 40%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-7-(methylamino)phenothiazin-5-ium chloride

A232505-25G A232505-5G
77877-777EA 166 USD
77877-777 77877-778
Azure C 40%
Azure C

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClN₃S
MW: 277.78 g/mol
MDL Number: MFCD00067677
CAS Number: 531-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR