Diethylamine 2-hydroxybenzoate ≥97%

Supplier: Ambeed
Warning

A232281-1G A232281-250MG A232281-5G
77600-864EA 35.88 USD
77600-864 77600-866 77626-226
Diethylamine 2-hydroxybenzoate ≥97%
Diethylamine 2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₃
MW: 211.26 g/mol
MDL Number: MFCD00045805
CAS Number: 4419-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR