trans-4-(Aminomethyl)cyclohexan-1-amine ≥95%

Supplier: Ambeed
Danger

A2320204-1G A2320204-25G
77877-775EA 13 USD
77877-775 77877-776
trans-4-(Aminomethyl)cyclohexan-1-amine ≥95%
trans-4-(Aminomethyl)cyclohexan-1-amine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂
CAS Number: 201211-54-3
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR