2,6-Difluoro-3,5-dipyridineboronic acid pinacol ester ≥96%

Supplier: Ambeed
Danger

Synonyms: 2,6-Difluoro-3,5-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A231764-1G A231764-250MG A231764-5G
77612-260EA 130.81 USD
77612-260 77612-262 77612-264
2,6-Difluoro-3,5-dipyridineboronic acid pinacol ester ≥96%
2,6-Difluoro-3,5-dipyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₅B₂F₂NO₄
MW: 367.01 g/mol
MDL Number: MFCD12913982
CAS Number: 1218789-90-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR