1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-2-(5-(1-(6-((2,5-dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-5-sulfoindolin-2-ylidene)penta-1,3-dien-1-yl)-3,3-dimethyl-3H-indol-1-ium-5-sulfonate ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cy 5
A231527-100MG
A231527-250MG
77748-393EA
567
USD
77748-393
77748-394
1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-2-(5-(1-(6-((2,5-dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-5-sulfoindolin-2-ylidene)penta-1,3-dien-1-yl)-3,3-dimethyl-3H-indol-1-ium-5-sulfonate ≥97%
1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-2-(5-(1-(6-((2,5-dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-5-sulfoindolin-2-ylidene)penta-1,3-dien-1-yl)-3,3-dimethyl-3H-indol-1-ium-5-sulfonate
Precaución: for Research Use Only
Formula:
C₄₅H₅₂N₄O₁₄S₂ MW: 937.04 g/mol |
MDL Number:
MFCD28978196 CAS Number: 146368-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...