(2R,5R,Z)-3-(2-Hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid ≥98%

Supplier: Ambeed
Danger

Synonyms: Clavulanic acid

A231352-10MG A231352-1MG A231352-5MG
77600-852EA 60.48 USD
77600-852 77600-854 77600-856
(2R,5R,Z)-3-(2-Hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid ≥98%
(2R,5R,Z)-3-(2-Hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₅
MW: 199.16 g/mol
MDL Number: MFCD00867004
CAS Number: 58001-44-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR