Theaflavin-3-gallate ≥98%

Supplier: Ambeed
Warning

A231018-10MG A231018-100MG A231018-50MG A231018-5MG
77612-248EA 216.3 USD
77612-248 77892-386 77892-387 77892-388
Theaflavin-3-gallate ≥98%
Theaflavin-3-gallate

Precaución: for Research Use Only

Formula: C₃₆H₂₈O₁₆
MW: 716.6 g/mol
MDL Number: MFCD03427508
CAS Number: 30462-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR