Tris(2,4-dimethylphenyl)phosphine ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A231014-1G A231014-5G
77683-672EA 14 USD
77683-672 77683-673
Tris(2,4-dimethylphenyl)phosphine ≥97%, Ambeed.Inc
Tris(2,4-dimethylphenyl)phosphine

Precaución: For research use only.

Formula: C₂₄H₂₇P
MW: 346.4450 g/mol
MDL Number: MFCD09039068
CAS Number: 49676-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR