3,5-Dichloro-4-fluorophenylboronic acid pinacol ester ≥97%

Supplier: Ambeed
Warning

Synonyms: 2-(3,5-Dichloro-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A230852-100MG A230852-1G A230852-250MG A230852-5G
77885-583EA 22 USD
77885-583 77885-584 77885-585 77885-586
3,5-Dichloro-4-fluorophenylboronic acid pinacol ester ≥97%
3,5-Dichloro-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₄BCl₂FO₂
MDL Number: MFCD15143599
CAS Number: 1092485-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR