N-Methyl-3-(1-methyl-1H-imidazol-4-yl)-4-((4-(trifluoromethyl)benzyl)amino)benzenesulfonamide ≥98%

Supplier: Ambeed
Warning

Synonyms: IK-930

A2305270-10MG A2305270-5MG
77626-214EA 43.26 USD
77626-214 77626-216
N-Methyl-3-(1-methyl-1H-imidazol-4-yl)-4-((4-(trifluoromethyl)benzyl)amino)benzenesulfonamide ≥98%
N-Methyl-3-(1-methyl-1H-imidazol-4-yl)-4-((4-(trifluoromethyl)benzyl)amino)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₉H₁₉F₃N₄O₂S
MW: 424.44 g/mol
MDL Number: MFCD34755308
CAS Number: 2563892-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR