8-Bromo-5-chloroquinoline ≥95%

Supplier: Ambeed
Warning

Synonyms: 8-Bromo-5-chloro-1-azanaphthalene

A230466-1G A230466-250MG A230466-5G
77593-494EA 59.45 USD
77593-494 77872-169 77872-170
8-Bromo-5-chloroquinoline ≥95%
8-Bromo-5-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MDL Number: MFCD13193679
CAS Number: 1154741-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR