Salvianolic acid C 99+%

Supplier: Ambeed
Warning

A230245-1MG A230245-5MG A230245-10MG
77612-242EA 17.51 USD
77612-242 77612-244 77612-240
Salvianolic acid C 99+%
Salvianolic acid C

Precaución: for Research Use Only

Formula: C₂₆H₂₀O₁₀
MDL Number: MFCD16660675
CAS Number: 115841-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR