5-Bromo-3-fluoropicolinaldehyde ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A230114-100MG A230114-250MG
77683-669EA 36 USD
77683-669 77683-670
5-Bromo-3-fluoropicolinaldehyde ≥98%, Ambeed.Inc
5-Bromo-3-fluoropicolinaldehyde

Precaución: For research use only.

Formula: C₆H₃BrFNO
MDL Number: MFCD14155977
CAS Number: 669066-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR