8-Methoxy-[1,3]dioxolo[4',5':4,5]benzo[1,2,3-cd]benzo[f]indol-5(6H)-one ≥97%

Supplier: Ambeed
Danger

Synonyms: Aristolactam I

A230023-1MG A230023-10MG A230023-5MG
77615-914EA 47.38 USD
77615-914 77746-341 77746-342
8-Methoxy-[1,3]dioxolo[4',5':4,5]benzo[1,2,3-cd]benzo[f]indol-5(6H)-one ≥97%
8-Methoxy-[1,3]dioxolo[4',5':4,5]benzo[1,2,3-cd]benzo[f]indol-5(6H)-one

Precaución: for Research Use Only

Formula: C₁₇H₁₁NO₄
MW: 293.27 g/mol
MDL Number: MFCD00797313
CAS Number: 13395-02-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR