Theophylline-7-acetic acid 99+%

Supplier: Ambeed
Warning

Synonyms: 2-(1,3-Dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)acetic acid

A229005-100G A229005-500G A229005-25G
77612-228EA 60.77 USD
77612-228 77672-030 77883-600
Theophylline-7-acetic acid 99+%
Theophylline-7-acetic acid

Precaución: for Research Use Only

Formula: C₉H₁₀N₄O₄
MDL Number: MFCD00022832
CAS Number: 652-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR