Ir(mppy)3-(OC-6-22)- 98+%

Supplier: Ambeed
Warning

A2289423-1G A2289423-250MG
77626-206EA 483.07 USD
77626-206 77877-752
Ir(mppy)3-(OC-6-22)- 98+%
Ir(mppy)3-(OC-6-22)-

Precaución: for Research Use Only

Formula: C₃₆H₃₀IrN₃
MDL Number: MFCD09038722
CAS Number: 149005-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR