Bis(2,5-dioxopyrrolidin-1-yl) adipate ≥99%

Supplier: Ambeed
Warning

Synonyms: Di(N-succinimidyl)adipate

A2289074-5G A2289074-1G A2289074-250MG
77593-470EA 179.38 USD
77593-470 77598-344 77626-200
Bis(2,5-dioxopyrrolidin-1-yl) adipate ≥99%
Bis(2,5-dioxopyrrolidin-1-yl) adipate

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₈
MW: 340.28 g/mol
MDL Number: MFCD01318516
CAS Number: 59156-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR