2-(5-Bromonaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: Ambeed
Warning

A2289051-100MG A2289051-250MG
77626-194EA 128.75 USD
77626-194 77626-196
2-(5-Bromonaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(5-Bromonaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₁₈BBrO₂
MW: 333.03 g/mol
CAS Number: 1807534-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR