(4-Bromo-3-chloro-5-(methoxycarbonyl)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A2289048-250MG A2289048-100MG A2289048-1G
77626-192EA 183.34 USD
77626-192 77877-744 77877-745
(4-Bromo-3-chloro-5-(methoxycarbonyl)phenyl)boronic acid ≥95%
(4-Bromo-3-chloro-5-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₇BBrClO₄
MW: 293.31 g/mol
CAS Number: 2788844-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR