(2-(4-Ethylphenoxy)pyridin-3-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A2289036-1G A2289036-100MG A2289036-250MG A2289036-5G
77626-190EA 289.43 USD
77626-190 77877-741 77877-742 77877-743
(2-(4-Ethylphenoxy)pyridin-3-yl)boronic acid ≥95%
(2-(4-Ethylphenoxy)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₄BNO₃
CAS Number: 2089158-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR