5-Methoxy-L-tryptophan ≥95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(5-methoxy-1H-indol-3-yl)propanoic acid, H-Trp(5-OMe)-OH

A228826-100MG A228826-1G
77626-182EA 21.63 USD
77626-182 77626-184
5-Methoxy-L-tryptophan ≥95%
5-Methoxy-L-tryptophan

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₃
MW: 234.251 g/mol
MDL Number: MFCD01074513
CAS Number: 25197-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR